Läs mer
Linaclotide, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Linaclotide |
| CAS | 851199-59-2 |
| Molekylformel | C59 H79 N15 O21 S6 |
| Kvantitet | 100 mg |
| Synonym | L-Tyrosine, L-cysteinyl-L-cysteinyl-L-α-glutamyl-L-tyrosyl-L-cysteinyl-L-cysteinyl-L-asparaginyl-L-prolyl-L-alanyl-L-cysteinyl-L-threonylglycyl-L-cysteinyl-, cyclic (1→6),(2→10),(5→13)-tris(disulfide), Linaclotide, Linzess |
| InChI formel | InChI=1S/C59H79N15O21S6/c1-26-47(82)69-41-25-101-99-22-38-52(87)65-33(13-14-45(80)81)49(84)66-34(16-28-5-9-30(76)10-6-28)50(85)71-40(54(89)72-39(23-97-96-20-32(60)48(83)70-38)53(88)67-35(18-43(61)78)58(93)74-15-3-4-42(74)56(91)63-26)24-100-98-21-37(64-44(79)19-62-57(92)46(27(2)75)73-55(41)90)51(86)68-36(59(94)95)17-29-7-11-31(77)12-8-29/h5-12,26-27,32-42,46,75-77H,3-4,13-25,60H2,1-2H3,(H2,61,78)(H,62,92)(H,63,91)(H,64,79)(H,65,87)(H,66,84)(H,67,88)(H,68,86)(H,69,82)(H,70,83)(H,71,85)(H,72,89)(H,73,90)(H,80,81)(H,94,95)/t26-,27?,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,46-/m0/s1 |
| LEDER | C[C@@H](O)[C@@H]1NC(=O)[C@@H]2CSSC[C@@H]3NC(=O)[C@@H](N)CSSC[C@H](NC(=O)[C@H](CSSC[C@H](NC(=O)CNC1=O)C(=O)N[C@@H](Cc4ccc(O)cc4)C(=O)O)NC(=O)[C@H](Cc5ccc(O)cc5)NC(=O)[C@H](CCC(=O)O)NC3=O)C(=O)N[C@@H](CC(=O)N)C(=O)N6CCC[C@H]6C(=O)N[C@@H](C)C(=O)N2 |
| Molekylvikt (g/mol) | 1526.74 |
| Formel vikt | 1525.3899 |