Läs mer
Linagliptin Methyldimer (>85%), TRC
Produktkod. 30133562
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133562 1 mg
30010152 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133562 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL4659101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1418133-47-7
Molekylformel C50H56N16O4
Molekylvikt (g/mol) 945.08
Synonym 8-[(3R)-3-Amino-1-piperidinyl]-1-[[4-[[2-[[8-[(3R)-3-amino-1-piperidinyl]-7-(2-butyn-1-yl)-2,3,6,7-tetrahydro-3-methyl-2,6-dioxo-1H-purin-1-yl]methyl]-1,4-dihydro-4-methyl-4-quinazolinyl]methyl]-2-quinazolinyl]methyl]-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1H-purine-2,6-dione, 8-((R)-3-Aminopiperidin-1-yl)-1-((4-((2-((8-((R)-3-aminopiperidin-1-yl)-7-(but-2-yn-1-yl)-3-methyl-2,6-dioxo-2,3,6,7-tetrahydro-1H-purin-1-yl)methyl)-4-methyl-1,4-dihydroquinazolin-4-yl)methyl)quinazolin-2-yl)methyl)-7-(but-2-yn-1-yl)-3-methyl-1H-purine-2,6(3H,7H)-dione, Linagliptin Dimer Impurity
LEDER c1cc2c(cc1)C(O)=C(C(=O)N2)C3=NS(=O)(=O)c4c(cccc4)N3
Specifikationer
Kemiskt namn eller material Linagliptin Methyldimer
Kvantitet 1 mg
InChI formel InChI=1S/C50H56N16O4/c1-6-8-24-63-40-42(56-46(63)61-22-14-16-31(51)27-61)59(4)48(69)65(44(40)67)29-38-53-35-20-12-10-18-33(35)37(55-38)26-50(3)34-19-11-13-21-36(34)54-39(58-50)30-66-45(68)41-43(60(5)49(66)70)57-47(64(41)25-9-7-2)62-23-15-17-32(52)28-62/h10-13,18-21,31-32H,14-17,22-30,51-52H2,1-5H3,(H,54,58)/t31-,32-,50?/m1/s1