Läs mer
Lincomycin 2-Phosphate, TRC
Produktkod. 17914275
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17914275 10 mg
17911155 5 mg
17903155 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17914275 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL46620510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 27480-30-4
Molekylformel C18H35N2O9PS
Molekylvikt (g/mol) 486.52
Synonym Methyl 6,8-dideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-D-erythro-α-D-galacto-Octopyranoside 2-(dihydrogen phosphate), Clindamycin Phosphate EP Impurity F
LEDER O[C@H]1[C@@H]([C@H](NC([C@H]2C[C@H](CCC)CN2C)=O)[C@H](O)C)O[C@H](SC)C(OP(O)(O)=O)C1O
Specifikationer
Kemiskt namn eller material Lincomycin 2-Phosphate
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C18H35N2O9PS/c1-5-6-10-7-11(20(3)8-10)17(24)19-12(9(2)21)15-13(22)14(23)16(18(28-15)31-4)29-30(25,26)27/h9-16,18,21-23H,5-8H2,1-4H3,(H,19,24)(H2,25,26,27)/t9-,10-,11+,12-,13-,14+,15-,16-,18-/m1/s1
Kvantitet 10 mg
Formel vikt 486.1801
Procent renhet >95%