missing translation for 'onlineSavingsMsg'
Learn More

Lincomycin Hydrochloride, TRC

Product Code. 30007405
Change view
Click to view available options
Kvantitet:
50 mg
1 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30007405 25 g
17912775 1 g
17974015 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007405 Supplier Toronto Research Chemicals Supplier No. TRCL46620025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 859-18-7
Molekylformel C18H34N2O6S.HCl
Molekylvikt (g/mol) 443
Synonym Lincomycin Hydrochloride, D-erythro-α-D-galacto-Octopyranoside, methyl 6,8-dideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-, hydrochloride (1:1), D-erythro-D-galacto-Octopyranoside, methyl 6,8-dideoxy-6-(1-methyl-4-propyl-L-2-pyrrolidinecarboxamido)-1-thio-, monohydrochloride, trans-α- (8CI), D-erythro-α-D-galacto-Octopyranoside, methyl 6,8-dideoxy-6-[[(1-methyl-4-propyl-2-pyrrolidinyl)carbonyl]amino]-1-thio-, monohydrochloride, (2S-trans)-, D-erythro-α-D-galacto-Octopyranoside, methyl 6,8-dideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-, monohydrochloride (9CI), Lincomycin hydrochloride (7CI), Lincocin, Lincogap, Lincomix, NSC 70731, U 10149a
IUPAC-namn (2S,4R)-N-[(1R,2R)-2-hydroxy-1-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride
LEDER Cl.CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@@H](C)O)[C@H]2O[C@H](SC)[C@H](O)[C@@H](O)[C@H]2O

Specifications

Kemiskt namn eller material Lincomycin Hydrochloride
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C18H34N2O6S.ClH/c1-5-6-10-7-11(20(3)8-10)17(25)19-12(9(2)21)16-14(23)13(22)15(24)18(26-16)27-4;/h9-16,18,21-24H,5-8H2,1-4H3,(H,19,25);1H/t9-,10-,11+,12-,13+,14-,15-,16-,18-;/m1./s1
Kvantitet 25 g
Formel vikt 442.1904
Procent renhet >95%
Frakt skick Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.