Läs mer
Linolenic-1-13C Acid, TRC
Produktkod. 30477196
Change view
Kvantitet:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30477196 25 mg
30477639 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30477196 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL46765325MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 224568-13-2
Molekylformel C1713CH30O2
Molekylvikt (g/mol) 279.42
Synonym (9Z,12Z,15Z)-9,12,15-Octadecatrienoic-1-13C Acid, (Z,Z,Z)-Octadeca-9,12,15-trienoic-1-13C Acid, (all-Z)-9,12,15-Octadecatrienoic-1-13C Acid, 9,12,15-all-cis-Octadecatrienoic-1-13C Acid, all-cis-9,12,15-Octadecatrienoic-1-13C Acid, cis-∆9,12,15-Octadecatrienoic-1-13C acid, α-Linolenic-1-13C Acid
IUPAC-namn (9Z,12Z,15Z)-octadeca-9,12,15-trienoic-1-13C acid
LEDER O[13C](CCCCCCC/C=C\C/C=C\C/C=C\CC)=O
Specifikationer
Kemiskt namn eller material Linolenic-1-13C Acid
Kvantitet 25 mg
InChI formel InChI=1S/C18H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h3-4,6-7,9-10H,2,5,8,11-17H2,1H3,(H,19,20)/b4-3-,7-6-,10-9-/i18+1
Formel vikt 279.23