Läs mer
Linolenic Acid Ethyl Ester, TRC
Produktkod. 30138641
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138641 10 g
30007713 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138641 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL46764510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1191-41-9
Molekylformel C20 H34 O2
Molekylvikt (g/mol) 306.48
Synonym (Z,Z,Z)-9,12,15-Octadecatrienoic acid ethyl ester, Ethyl linolenate
IUPAC-namn ethyl (9Z,12Z,15Z)-octadeca-9,12,15-trienoate
LEDER CCOC(=O)CCCCCCC\C=C/C\C=C/C\C=C/CC
Specifikationer
Kemiskt namn eller material Linolenic acid ethyl ester
Anteckningar om renhetsgrad GC
Kvantitet 10 g
InChI formel InChI=1S/C20H34O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h5-6,8-9,11-12H,3-4,7,10,13-19H2,1-2H3/b6-5-,9-8-,12-11-
Formel vikt 306.2559
Procent renhet >95%