Läs mer
Linoleoyl Chloride, TRC
Produktkod. 30465054
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30465054 1 g
1 options
Produktkod. 30465054 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL4676701G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Linoleoyl Chloride
CAS 7459-33-8
Molekylformel C18 H31 Cl O
Kvantitet 1 g
Synonym (9Z,12Z)-9,12-Octadecadienoyl Chloride, (Z,Z)-9,12-Octadecadienoyl Chloride, (Z,Z)-Octadeca-9,12-dienoyl Chloride, Linoleic Acid Chloride
InChI formel InChI=1S/C18H31ClO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h6-7,9-10H,2-5,8,11-17H2,1H3/b7-6-,10-9-
LEDER CCCCC\C=C/C\C=C/CCCCCCCC(=O)Cl
IUPAC-namn (9Z,12Z)-octadeca-9,12-dienoyl chloride
Molekylvikt (g/mol) 298.89
Formel vikt 298.2063