Learn More
LINS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558450
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SRVFIEQDDD MLEAAKASLG IYLTLTRGCE ATESLTQGKE MWDHHTHENG YNPHCIFLFF LKNIGFDSTV LLDFLISSET CFLEYFVRYL KLLQKDWDN Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 81%.
LINS Polyclonal antibody specifically detects LINS in Human samples. It is validated for Immunohistochemistry.Specifications
LINS | |
Polyclonal | |
Unconjugated | |
LINS1 | |
lines homolog 1; LINS; LINS1; MRT27; Protein Lines homolog 1; WINS1; WINS1 protein with Drosophila Lines (Lin) homologous domain; wnt-signaling molecule Lines homolog 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55180 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NG48 | |
LINS1 | |
Recombinant protein corresponding to Human LINS | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only