Läs mer
Lipstatin, TRC
Produktkod. 30475733
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30475733 10 mg
30465848 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30475733 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL46880010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 96829-59-3
Molekylformel C29H49NO5
Molekylvikt (g/mol) 491.7
Synonym N-Formyl-L-leucine (1S,3Z,6Z)-1-[[(2S,3S)-3-Hexyl-4-oxo-2-oxetanyl]methyl]-3,6- dodecadien-1-yl Ester, N-Formyl-L-leucine [2S-[2α(1R*,3Z,6Z),3β]]-1- [(3-Hexyl-4-oxo-2-oxetanyl)methyl]-3,6-dodecadienyl Ester, (-)-Lipstatin
LEDER CCCCCC[C@H]1[C@@H](OC1=O)C[C@H](C/C=C\C/C=C\CCCCC)OC(=O)[C@H](CC(C)C)NC=O
Specifikationer
Kemiskt namn eller material Lipstatin
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C29H49NO5/c1-5-7-9-11-12-13-14-15-16-18-24(34-29(33)26(30-22-31)20-23(3)4)21-27-25(28(32)35-27)19-17-10-8-6-2/h12-13,15-16,22-27H,5-11,14,17-21H2,1-4H3,(H,30,31)/b13-12-,16-15-/t24-,25-,26-,27-/m0/s1
Formel vikt 491.3611
Procent renhet >95%