Läs mer
Lisadimate (>85%), TRC
Produktkod. 30471666
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30471666 10 mg
30466960 50 mg
30478837 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30471666 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL46000010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 136-44-7
Molekylformel C10H13NO4
Molekylvikt (g/mol) 211.21
Synonym Glyceryl p-aminobenzoate, Escalol 106, Glycerol, 1-(p-aminobenzoate), 2,3-Dihydroxypropyl 4-aminobenzoate, 4-Aminobenzoic acid 1-glyceryl ester, NIPA GMPA
IUPAC-namn 2,3-dihydroxypropyl 4-aminobenzoate
LEDER Nc1ccc(cc1)C(=O)OCC(O)CO
Specifikationer
Kemiskt namn eller material Lisadimate
Kvantitet 10 mg
InChI formel InChI=1S/C10H13NO4/c11-8-3-1-7(2-4-8)10(14)15-6-9(13)5-12/h1-4,9,12-13H,5-6,11H2
Formel vikt 211.0845