Läs mer
Lisinopril Cyclohexyl Analogue, TRC
Produktkod. 30476922
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30476922 5 mg
1 options
Produktkod. 30476922 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL4690005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2S)-1-[(2S)-6-Amino-2-[[(1S)-1-carboxy-3-cyclohexylpropyl]amino]hexanoyl]-pyrrolidine-2-carboxylic Acid
CAS 1132650-67-9
Molekylformel C21 H37 N3 O5
Kvantitet 5 mg
Synonym Lisinopril Imp. F (EP), (2S)-1-[(2S)-6-Amino-2-[[(1S)-1-carboxy-3-cyclohexylpropyl]amino]hexanoyl]-pyrrolidine-2-carboxylic Acid, Lisinopril Cyclohexyl Analogue, N2-[(1S)-1-carboxy-3-cyclohexylpropyl]-L-lysyl-L-Proline
InChI formel InChI=1S/C21H37N3O5/c22-13-5-4-9-16(19(25)24-14-6-10-18(24)21(28)29)23-17(20(26)27)12-11-15-7-2-1-3-8-15/h15-18,23H,1-14,22H2,(H,26,27)(H,28,29)/t16-,17-,18-/m0/s1
LEDER NCCCC[C@H](N[C@@H](CCC1CCCCC1)C(=O)O)C(=O)N2CCC[C@H]2C(=O)O
IUPAC-namn (2S)-1-[(2S)-6-amino-2-[[(1S)-1-carboxy-3-cyclohexylpropyl]amino]hexanoyl]pyrrolidine-2-carboxylic acid
Molekylvikt (g/mol) 411.54
Formel vikt 411.2733