missing translation for 'onlineSavingsMsg'
Learn More

Lithium Bromide Solution, Ultra-Pure Additive, SPEX CertiPrep™

Catalog Number p-4384784
Click to view available options
Quantity:
15 mL
10 x 15 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
7550-35-8
BrLi
86.844
AMXOYNBUYSYVKV-UHFFFAOYSA-M
lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide
82050
CHEBI:63042
lithium;bromide
[Li+].[Br-]
This item is not returnable. View return policy
7550-35-8
BrLi
86.844
AMXOYNBUYSYVKV-UHFFFAOYSA-M
lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide
82050
CHEBI:63042
lithium;bromide
[Li+].[Br-]

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Ultra Pure Additives
  • Lithium Bromide Solution
  • 15 mL Package Size

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

Quantity 15 mL
Grade Ultrapure
Packaging Triple-leached LDPE bottle
Chemical Name or Material Lithium Bromide Solution, Additive Ultra Pure
compliance-icons
Product Identifier
  • fusion flux-ultra pure additive
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : lithium bromide

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'