missing translation for 'onlineSavingsMsg'
Learn More

Lithium Fluoride, Supelco™

Product Code. 15847380
Change view
Click to view available options
Kvantitet:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet Unit Size
15847380 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ100 Graded - For Non-regulated Laboratory Applications
Product Code. 15847380 Supplier Supelco Supplier No. 1056860050
 Begränsad artikel
Beräknad leverans: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ100 Graded - For Non-regulated Laboratory Applications

99.99 Suprapur™

We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This enabling product has been enhanced for energy efficiency.

TRUSTED_SUSTAINABILITY

Specifications

Kokpunkt 1676°C/1013 hPa
Kemiskt namn eller material Lithium Fluoride
Smältpunkt 870°C
CAS 7789-24-4
Kvantitet 50 g
MDL-nummer MFCD00011090
Synonym Lithium fluoride, Lithium fluoride
Molekylvikt (g/mol) 25.94
Kvalitet Research
Densitet 2.64 g/cm3 at 20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.