missing translation for 'onlineSavingsMsg'
Learn More

Lithium Iodide Solution, Ultra-Pure Additive, SPEX CertiPrep™

Catalog Number 15206376
Click to view available options
Quantity:
15 mL
10 x 15 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Ultra Pure Additives
  • Lithium Iodide Solution
  • 15 mL Package Size

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

Quantity 15 mL
Grade Ultrapure
Packaging Triple-leached LDPE bottle
Chemical Name or Material Lithium Iodide Solution, Additive Ultra Pure

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'