missing translation for 'onlineSavingsMsg'
Learn More

Lithium Perchlorate, ACS Reagent, ≥95.0%, Honeywell™ Fluka™

Catalog Number p-7120524
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
7791-03-9
ClH6LiO7
160.43
MFCD00011079
JAWYRNYHJJDXHX-UHFFFAOYSA-M
This item is not returnable. View return policy
7791-03-9
ClH6LiO7
160.43
MFCD00011079
JAWYRNYHJJDXHX-UHFFFAOYSA-M

ACS Reagent,≥95.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 236°C
Color White
pH 6.0 to 9.5 (at 106.4g/L 25°C)
Quantity 5 g
Linear Formula LiClO4
UN Number UN1481
Synonym lithium perchlorate, perchloric acid, lithium salt, lithiumperchlorate, unii-q86se98c9c, lithium 1+ ion perchlorate, lithium perchlorate, anhydrous, lithium cloricum, lithium per chlorate, liclo4, perchloric acid, lithium salt 1:1
SMILES [Li+].[O-]Cl(=O)(=O)=O
IUPAC Name lithium;perchlorate
PubChem CID 23665649
Formula Weight 106.39g/mol
Percent Purity ≥95.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Granules
Density 2.420g/cu.cm
Chemical Name or Material Lithium Perchlorate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'