missing translation for 'onlineSavingsMsg'
Learn More

Lithium perchlorate trihydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog Number p-7048840
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
13453-78-6
ClH6LiO7
160.431
MFCD00149765
JAWYRNYHJJDXHX-UHFFFAOYSA-M
lithium perchlorate trihydrate, perchloric acid, lithium salt, trihydrate, unii-jyk745743v, acmc-1c1zx, lithium perchlorate hydrate, lithiumperchlorate trihydrate, ksc491a4p, lithium pechlorate trihydrate, lithiumperchloratetrihydrate, lithotab trihydrate perchlorate ion
23708903
lithium;perchlorate;trihydrate
[Li+].O.O.O.[O-]Cl(=O)(=O)=O
This item is not returnable. View return policy
13453-78-6
ClH6LiO7
160.431
MFCD00149765
JAWYRNYHJJDXHX-UHFFFAOYSA-M
lithium perchlorate trihydrate, perchloric acid, lithium salt, trihydrate, unii-jyk745743v, acmc-1c1zx, lithium perchlorate hydrate, lithiumperchlorate trihydrate, ksc491a4p, lithium pechlorate trihydrate, lithiumperchloratetrihydrate, lithotab trihydrate perchlorate ion
23708903
lithium;perchlorate;trihydrate
[Li+].O.O.O.[O-]Cl(=O)(=O)=O

It is used as an oxygen source for chemical oxygen generators, as it liberates oxygen at high temperatures. Excellent solubility of lithium perchlorate permits its use in Diels-Alder reactions. It is also employed in lithium-ion batteries as an electrolyte, and act as chaotropic agent to denature proteins. It is used in Baylis-Hillman reaction as a catalyst in the coupling of alpha,beta-unsaturated carbonyl compounds with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Crystalline
Quantity 250 g
Linear Formula LiClO4·3H2O
UN Number UN1481
Merck Index 14,5539
Solubility Information Highly soluble in water,methanol,ethanol,ethyl acetate,diethyl ether,and acetone.
Formula Weight 160.45 (106.40 Anhydrous)
Grade Reagent
Odor Odorless
Density 1.84 g/mL
Chemical Name or Material Lithium perchlorate trihydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'