missing translation for 'onlineSavingsMsg'
Learn More

Lithium tetrakis(pentafluorophenyl)borate ethyl etherate, Thermo Scientific Chemicals

Catalog Number p-7077738
Click to view available options
Quantity:
100 mg
1 g
This item is not returnable. View return policy
371162-53-7
2·5 C4H10O
871.28
KPLZKJQZPFREPG-UHFFFAOYSA-N
lithium tetrakis pentafluorophenyl borate ethyl etherate, lithium tetrakis pentafluorophenyl borate, tetrakis pentafluorophenyl boron lithium ethyl etherate, lithium tetrakis pentafluorophenyl borate-ethyl ether complex, lithium tetrakis pentafluorophenyl borate ethyl etherate 2:2:5, lithium tetrakis pentafluorophenyl borate diethylether complex 1:2.5
23701484
lithium;ethoxyethane;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
[Li+].[B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.CCOCC
This item is not returnable. View return policy
371162-53-7
2·5 C4H10O
871.28
KPLZKJQZPFREPG-UHFFFAOYSA-N
lithium tetrakis pentafluorophenyl borate ethyl etherate, lithium tetrakis pentafluorophenyl borate, tetrakis pentafluorophenyl boron lithium ethyl etherate, lithium tetrakis pentafluorophenyl borate-ethyl ether complex, lithium tetrakis pentafluorophenyl borate ethyl etherate 2:2:5, lithium tetrakis pentafluorophenyl borate diethylether complex 1:2.5
23701484
lithium;ethoxyethane;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
[Li+].[B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.CCOCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 117.0°C to 122.0°C
Quantity 100 mg
Formula Weight 871.28
Chemical Name or Material Lithium tetrakis(pentafluorophenyl)borate ethyl etherate
compliance-icons
Product Identifier
  • Lithium tetrakis(pentafluorophenyl)borate ethyl etherate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Lithium tetrakis(pentafluorophenyl)borate ethyl etherate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'