Learn More
Shamrock™ Litho Color Coded Dot Labels
Offered in a number of colors and diameters to fit your needs. Shamrock™ Color Coded Dot Labels offer multiple opportunities for color coding in the laboratory - test, physician or department.
Brand:  Shamrock™ SDL-25R-3
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11746975
  • 125.00 SEK / Pack of 1000
Estimated Shipment: 07-06-2024
to see stock.
Description
Description
  • Resistant to water, oil, and acid
  • Temperature range of -20 to +121°C
  • For autoclave, freezer, refrigerator and oven
  • Universal marking with ballpoint pen, paper pencil and felt
Specifications
Specifications
Permanent
Roll
Color Coded Dot Label
Green
Litho
6.5 mm