Läs mer
Lithocholic Acid 3-O-Glucuronide, TRC
Produktkod. 30137752
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137752 5 mg
30135801 10 mg
30004274 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137752 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL4691905MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 75239-91-7
Molekylformel C30H48O9
Molekylvikt (g/mol) 552.7
Synonym (3α,5β)-23-Carboxy-24-norcholan-3-yl β-D-Glucopyranosiduronic Acid, Lithocholic Acid 3-Glucuronide;
LEDER [H][C@@]12C[C@H](O[C@@H]3O[C@H](C(O)=O)[C@@H](O)C(O)[C@H]3O)CC[C@]1(C)[C@]4([H])[C@@]([C@@]5([H])[C@]([C@@](CC5)([H])[C@H](C)CCC(O)=O)(C)CC4)([H])CC2
Specifikationer
Kemiskt namn eller material Lithocholic Acid 3-O-Glucuronide
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C30H48O9/c1-15(4-9-22(31)32)19-7-8-20-18-6-5-16-14-17(10-12-29(16,2)21(18)11-13-30(19,20)3)38-28-25(35)23(33)24(34)26(39-28)27(36)37/h15-21,23-26,28,33-35H,4-14H2,1-3H3,(H,31,32)(H,36,37)/t15-,16-,17+,18+,19-,20+,21+,23+,24+,25-,26+,28-,29+,30-/m1/s1
Procent renhet >95%