Läs mer
Lithocholic Sulfate Disodium Salt, TRC
Produktkod. 30138307
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138307 50 mg
30003504 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138307 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL46918550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 64936-81-8
Molekylformel C24H38Na2O6S
Molekylvikt (g/mol) 500.6
Synonym 3-(Sulfooxy)-(3alpha,5beta)-cholan-24-oic Acid, Disodium Lithocholic Acid Sulfate;
IUPAC-namn disodium;(4R)-4-[(3R,5R,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-3-sulfonatooxy-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
LEDER [Na+].[Na+].C[C@H](CCC(=O)[O-])[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@@H](CC[C@]4(C)[C@H]3CC[C@]12C)OS(=O)(=O)[O-]
Specifikationer
Kemiskt namn eller material Lithocholic Sulfate Disodium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C24H40O6S.2Na/c1-15(4-9-22(25)26)19-7-8-20-18-6-5-16-14-17(30-31(27,28)29)10-12-23(16,2)21(18)11-13-24(19,20)3;;/h15-21H,4-14H2,1-3H3,(H,25,26)(H,27,28,29);;/q;2*+1/p-2/t15-,16-,17-,18+,19-,20+,21+,23+,24-;;/m1../s1
Formel vikt 500.2184
Procent renhet >95%