Learn More
LLGL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554637
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: FERFSLSARN ITEPLCSLDI NWPRDATQAS YRIRESPKLS QANGTPSILL APQSLDGSPD PAHSMGPDTP EPPEAALSPM SIDSATSADT TLDTTGDVTV EDVKDFLGSS EESEKNLRNL AEDE Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 72%.
LLGL1 Polyclonal antibody specifically detects LLGL1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
LLGL1 | |
Polyclonal | |
Unconjugated | |
LLGL1 | |
AI325176; DLG4; HUGL; Hugl protein; HUGL1; Hugl-1; Human homolog to the D-lgl gene protein; lethal giant larvae homolog 1; lethal giant larvae homolog 1 (Drosophila); lethal giant larvae homolog 1, scribble cell polarity complex component; lethal(2) giant larvae protein homolog 1; Lgl1; LLGL; Llgl1; LLGL1, scribble cell polarity complex component; Llglh; Mgl1; Mgl-1; Mlgl; Rgl1; Rgl-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3996 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15334 | |
LLGL1 | |
Recombinant protein corresponding to Human LLGL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only