Läs mer
Tocris Bioscience™ LM11A 31 dihydrochloride
GREENER_CHOICE
Nonpeptide p75NTR ligand; neuroprotective
Brand:  Tocris Bioscience™ 5046/10
Encompass
Produktkod. 15746406
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
LM11A 31 dihydrochloride is a nonpeptide p75NTR ligand and nerve growth factor (NGF) antagonist. LM11A 31 blocks p75-mediated cell death, inhibits proNGF induced cell death (IC50 = 1-10 nM), increases proliferation and survival of hippocampal neural progenitors and inhibits Aβ-induced cell death (EC50 = 20 nM). In animal models of Alzheimer's disease LM11A 31 prevents and reverses atrophy of cholinergic neurites and reverses Alzheimer's pathology; in models of Huntington's disease LM11A 31 increases survival, reduces the formation of aggregates of mutant huntingtin and neuronal degeneration. Also promotes functional recovery in a mouse model of spinal cord injury. Orally available and brain penetrant.
Specifikationer
Specifikationer
95%
1243259-19-9
10 mg
(2 S,3 S)-2-Amino-3-methyl-N-[2-(4-morpholinyl)ethyl]pentanamide dihydrochloride
Desiccate at RT
Produktförslag
Produktförslag
Filmer
SDS
Dokument
Dokument
Certifikat
Kampanjer
Kampanjer