missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ LMO4 Recombinant Protein
Brand: Abnova™ H00008543-P01.10ug
Additional Details : Weight : 0.00010kg
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH17673 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
43.89 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
MVNPGSSSQPPPVTAGSLSWKRCAGCGGKIADRFLLYAMDSYWHSRCLKCSCCQAQLGDIGTSCYTKSGMILCRNDYIRLFGNSGACSACGQSIPASELVMRAQGNVYHLKCFTCSTCRNRLVPGDRFHYINGSLFCEHDRPTALINGHLNSLQSNPLLPDQKVC | |
LMO4 | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
8543 | |
LMO4 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LMO4 | |
Wheat Germ (in vitro) | |
GST |