missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ LMO6 (Human) Recombinant Protein
Human LMO6 full-length ORF ( AAH16856, 1 a.a. - 615 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004007-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MFARGSRRRRSGRAPPEAEDPDRGQPCNSCREQCPGFLLHGWRKICQHCKCPREEHAVHAVPVDLERIMCRLISDFQRHSISDDDSGCASEEYAWVPPGLKPEQVYQFFSCLPEDKVPYVNSPGEKYRIKQLLHQLPPHDSEAQYCTALEEEEKKELRAFSQQRKRENLGRGIVRIFPVTITGAICEECGKQIGGGDIAVFASRAGLGACWHPQCFVCTTCQELLVDLIYFYHVGKVYCGRHHAECLRPRCQACDEIIFSPECTEAEGRHWHMDHFCCFECEASLGGQRYVMRQSRPHCCACYEARHAEYCDGCGEHIGLDQGQMAYEGQHWHASDRCFCCSRCGRALLGRPFLPRRGLIFCSRACSLGSEPTAPGPSRRSWSAGPVTAPLAASTASFSAVKGASETTTKGTSTELAPATGPEEPSRFLRGAPHRHSMPELGLRSVPEPPPESPGQPNLRPDDSAFGRQSTPRVSFRDPLVSEGGPRRTLSAPPAQRRRPRSPPPRAPSRRRHHHHNHHHHHNRHPSRRRHYQCDAGSGSDSESCSSSPSSSSSESSEDDGFFLGERIPLPPHLCRPMPAQDTAMETFNSPSLSLPRDSRAGMPRQARDKNCIVA
Specifications
AAH16856 | |
prickle homolog 3 (Drosophila) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PRICKLE3 | |
GST |
4007 | |
Wheat germ expression system | |
25 μg | |
LMO6 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |