missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LNPEP Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55154-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to LNPEP.
The protein encoded by this gene is a member of the SEC24 subfamily of the SEC23/SEC24 family, which is involved in vesicle trafficking. The encoded protein has similarity to yeast Sec24p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. This gene product is implicated in the shaping of the vesicle, and also in cargo selection and concentration.Specifications
LNPEP | |
Polyclonal | |
Unconjugated | |
LNPEP | |
CAP, Cystinyl aminopeptidase, IRAP, Leucyl cystinyl aminopeptidase, leucyl/cystinyl aminopeptidase, LNPEP, OTase, Oxytocinase, P LAP, PLAP, VP165 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4012 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.67 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UIQ6 | |
LNPEP | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |