missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LNPEP Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | LNPEP |
---|---|
Concentration | 0.67 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16876625
|
Proteintech
55154-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 03-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16866625
|
Proteintech
55154-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 03-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody is specific to LNPEP.
The protein encoded by this gene is a member of the SEC24 subfamily of the SEC23/SEC24 family, which is involved in vesicle trafficking. The encoded protein has similarity to yeast Sec24p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. This gene product is implicated in the shaping of the vesicle, and also in cargo selection and concentration.Specifications
LNPEP | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9UIQ6 | |
4012 | |
Peptide | |
Primary | |
-20°C | |
LNPEP |
0.67 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CAP, Cystinyl aminopeptidase, IRAP, Leucyl cystinyl aminopeptidase, leucyl/cystinyl aminopeptidase, LNPEP, OTase, Oxytocinase, P LAP, PLAP, VP165 | |
LNPEP | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |