missing translation for 'onlineSavingsMsg'
Learn More

LOC84740 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10491-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16886903

  • 1236.20 SEK / 20µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene produces a long non-coding RNA that is overexpressed in tumor cells and may promote cancer cell metastasis.
Specifications

Specifications

LOC84740
0.48 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Liquid
AFAP1-AS1
LOC84740 Fusion Protein Ag0759
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human
ELISA
Unconjugated
AFAP1-AS1
hypothetical LOC84740, LOC84740
Rabbit
IgG
20 μL
-20°C
Primary
84740
Videos
SDS
Documents
Certificates
Special Offers

Special Offers