Läs mer
Loganic Acid, TRC
Produktkod. 30472888
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30472888 10 mg
1 options
Produktkod. 30472888 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL46940810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Loganic acid
CAS 22255-40-9
Molekylformel C16 H24 O10
Kvantitet 10 mg
Synonym Cyclopenta[c]pyran-4-carboxylic acid, 1-(β-D-glucopyranosyloxy)-1,4a,5,6,7,7a-hexahydro-6-hydroxy-7-methyl-, (1S,4aS,6S,7R,7aS)-, Cyclopenta[c]pyran-4-carboxylic acid, 1-(β-D-glucopyranosyloxy)-1,4a,5,6,7,7a-hexahydro-6-hydroxy-7-methyl-, [1S-(1α,4aα,6α,7α,7aα)]-, Cyclopenta[c]pyran-4-carboxylic acid, 1α-(β-D-glucopyranosyloxy)-1,4aα,5,6,7,7aα-hexahydro-6α-hydroxy-7α-methyl- (8CI), Loganic acid (6CI), (1S,4aS,6S,7R,7aS)-1-(β-D-Glucopyranosyloxy)-1,4a,5,6,7,7a-hexahydro-6-hydroxy-7-methylcyclopenta[c]pyran-4-carboxylic acid, Loganin acid
InChI formel InChI=1S/C16H24O10/c1-5-8(18)2-6-7(14(22)23)4-24-15(10(5)6)26-16-13(21)12(20)11(19)9(3-17)25-16/h4-6,8-13,15-21H,2-3H2,1H3,(H,22,23)/t5-,6+,8-,9+,10+,11+,12-,13+,15-,16-/m0/s1
LEDER C[C@H]1[C@@H](O)C[C@H]2[C@@H]1[C@H](O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)OC=C2C(=O)O
IUPAC-namn (1S,4aS,6S,7R,7aS)-6-hydroxy-7-methyl-1-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-1,4a,5,6,7,7a-hexahydrocyclopenta[c]pyran-4-carboxylic acid
Molekylvikt (g/mol) 376.36
Formel vikt 376.1369