Läs mer
Lomefloxacin Aspartate, TRC
Produktkod. 30468756
Change view
Kvantitet:
1 g
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30468756 1 g -
30467183 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30468756 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL4694101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 211690-33-4
Molekylformel C17 H19 F2 N3 O3 . C4 H7 N O4
Molekylvikt (g/mol) 484.45
Synonym L-Aspartic acid, 1-ethyl-6,8-difluoro-1,4-dihydro-7-(3-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylate (1:1), L-Aspartic acid, 1-ethyl-6,8-difluoro-1,4-dihydro-7-(3-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylate (9CI), 3-Quinolinecarboxylic acid, 1-ethyl-6,8-difluoro-1,3-dihydro-7-(3-methyl-1-piperazinyl)-4-oxo-, L-aspartate (1:1) (9CI), Lomefloxacin Aspartate, Lomefloxacin L-Aspartate
IUPAC-namn (2S)-2-aminobutanedioic acid;1-ethyl-6,8-difluoro-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid
LEDER CCN1C=C(C(=O)O)C(=O)c2cc(F)c(N3CCNC(C)C3)c(F)c12.N[C@@H](CC(=O)O)C(=O)O
Specifikationer
Kemiskt namn eller material Lomefloxacin Aspartate
Kvantitet 1 g
InChI formel InChI=1S/C17H19F2N3O3.C4H7NO4/c1-3-21-8-11(17(24)25)16(23)10-6-12(18)15(13(19)14(10)21)22-5-4-20-9(2)7-22;5-2(4(8)9)1-3(6)7/h6,8-9,20H,3-5,7H2,1-2H3,(H,24,25);2H,1,5H2,(H,6,7)(H,8,9)/t;2-/m.0/s1
Formel vikt 484.177