Läs mer
Lomifylline, TRC
Produktkod. 30469346
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30469346 50 mg
30479790 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30469346 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL47020050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 10226-54-7
Molekylformel C13 H18 N4 O3
Molekylvikt (g/mol) 278.31
Synonym 3,7-Dihydro-1,3-dimethyl-7-(5-oxohexyl)-1H-purine-2,6-dione, 7-(5-Oxohexyl)theophylline, 1,3-Dimethyl-7-(5-oxohexyl)xanthine, 7-(5-Oxohexyl)-1,3-dimethylxanthine, Cervilane
IUPAC-namn 1,3-dimethyl-7-(5-oxohexyl)purine-2,6-dione
LEDER CN1C(=O)N(C)c2ncn(CCCCC(=O)C)c2C1=O
Specifikationer
Kemiskt namn eller material Lomifylline
Kvantitet 50 mg
InChI formel InChI=1S/C13H18N4O3/c1-9(18)6-4-5-7-17-8-14-11-10(17)12(19)16(3)13(20)15(11)2/h8H,4-7H2,1-3H3
Formel vikt 278.1379