Learn More
Lomifylline, TRC
Product Code. 30479790
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479790 500 mg 500mg
30469346 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30479790 Supplier Toronto Research Chemicals Supplier No. TRCL470200500MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10226-54-7
Molecular Formula C13 H18 N4 O3
Molecular Weight (g/mol) 278.31
Synonym 3,7-Dihydro-1,3-dimethyl-7-(5-oxohexyl)-1H-purine-2,6-dione, 7-(5-Oxohexyl)theophylline, 1,3-Dimethyl-7-(5-oxohexyl)xanthine, 7-(5-Oxohexyl)-1,3-dimethylxanthine, Cervilane
IUPAC Name 1,3-dimethyl-7-(5-oxohexyl)purine-2,6-dione
SMILES CN1C(=O)N(C)c2ncn(CCCCC(=O)C)c2C1=O
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C13H18N4O3/c1-9(18)6-4-5-7-17-8-14-11-10(17)12(19)16(3)13(20)15(11)2/h8H,4-7H2,1-3H3
Formula Weight 278.1379
Chemical Name or Material Lomifylline