Learn More
Abnova™ LongTaq DNA Polymerase
LongTaq DNA polymerase catalyzes 5'-3' synthesis of DNA.
4610.00 SEK - 18355.00 SEK
Specifications
Content And Storage | Store at -20°C Guaranteed stable for 6 months when properly stored |
---|---|
For Use With (Application) | Standard PCR |
Concentration | 5 units/μL |
Storage Buffer | 20 mM Tris-HCl (pH 8.0), 1 mM DTT, 0.1 mM EDTA, 100 mM KCl, 0.5% Nonidet P40, 0.5% Tween 20 and 50% glycerol |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16052555
|
Abnova™
D0013.1000UNITS |
1000 units |
9215.00 SEK
1000 units |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16062555
|
Abnova™
D0013.2500UNITS |
2500 units |
18355.00 SEK
2500 units |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16072555
|
Abnova™
D0013.500UNITS |
500 units |
4610.00 SEK
500 units |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- The enzyme has been proved not having the 3′-5′ exonuclease activity.
- The enzyme has proven to have high amplification yield, be stable at high temparetaure.
Specifications
Store at -20°C Guaranteed stable for 6 months when properly stored | |
Standard PCR | |
5 units/μL | |
20 mM Tris-HCl (pH 8.0), 1 mM DTT, 0.1 mM EDTA, 100 mM KCl, 0.5% Nonidet P40, 0.5% Tween 20 and 50% glycerol |