Läs mer
Tocris Bioscience™ Lonidamine
GREENER_CHOICE
Produktkod. 18762564
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18762564 10 mg
18742674 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18762564 Leverantör Tocris Bioscience™ Leverantörsnummer 1646/10
Mitochondrial hexokinase inhibitor
Anticancer and antispermatogenic agent in vitro and in vivo. Inhibits cellular energy metabolism in some cells via inhibition of mitochondrial hexokinase. Also blocks CFTR Cl- channels in vitro.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 50264-69-2
Molekylformel C15H10Cl2N2O2
Specifikationer
Kemiskt namn eller material Lonidamine
Kvantitet 10 mg
Formel vikt 321.16
Renhet >98%
Molekylvikt (g/mol) 321.157