Learn More
Lonza™ BioWhittaker™ Antibiotics: Gentamicin Sulfate
Used as a broad spectrum bactericidal, with activity against gram negative and gram positive organisms including Pseudomonas. BioWhittaker™ Antibiotics: Gentamicin Sulfate is manufactured in accordance with cGMP regulations and cell culture tested to assure compatibility.
Brand: Lonza™ 17-528Z
Additional Details : Weight : 0.45360kg
Description
- Broad spectrum bactericida
- Stable over a wide range of pH and temperature
- Autoclavable
- Colorless to amber or yellow.
- cGMP compatibility
Specifications
15° to 30°C | |
Antibiotic | |
3.5 to 5.5 |
50 mg/mL | |
Crimp top vial | |
10 mL |