Learn More
Lonza™ BioWhittaker™ Antibiotics: Penicillin-Streptomycin Mixtures
Used as a broad spectrum bacteriostatic and bactericidal, with activity against gram negative and gram-positive organisms including yeast and mold. BioWhittaker™ Antibiotics: Penicillin-Streptomycin Mixtures are manufactured in accordance with cGMP regulations.
347.00 SEK - 6070.00 SEK
Specifications
Content And Storage -10° to -20°C
View More Specs
Products 6
Product Code Brand Quantity Product Type Price Quantity & Availability  
Product Code Brand Quantity Product Type Price Quantity & Availability  
11625150
View Documents Promotion Details
Lonza™
09-757F
500 mL Antibiotic This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11629990
View Documents Promotion Details
Lonza™
DE17-602E
100 mL Antibiotic This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11659990
View Documents Promotion Details
Lonza™
DE17-603E
100 mL Antibiotic This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11630001
View Documents Promotion Details
Lonza™
17-718R
25 x 4.5 mL Antibiotic-L-Glutamine This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11690001
View Documents Promotion Details
Lonza™
17-745E
100 mL Antibiotic-Antimycotic This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11600011
View Documents Promotion Details
Lonza™
17-745H
20 mL Antibiotic-Antimycotic This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • Suggested working concentration of Penicillin-Streptomycin is 100 units of Potassium Penicillin and 100 μg of Streptomycin Sulfate per 1 ml of culture media
  • Manufactured in accordance with cGMP regulations
  • Cell culture tested to assure compatibility.
  • 20mL vials have crimp-top closure
  • 4.5, 100 and 500mL bottles have screw-cap closure
Specifications
Specifications
-10° to -20°C