Learn More
Lonza™ BioWhittaker™ HEPES Buffer
Used in open vessels incubated under atmospheric conditions. BioWhittaker™ HEPES Buffer is manufactured in accordance with cGMP regulations.
Brand: Lonza™ BE17-737E
Additional Details : Weight : 0.16000kg
Description
- Used in open vessels incubated under atmospheric conditions
- Behaves as zwitterion
- Sodium chloride concentration of HEPES buffered medium has been reduced to maintain same Osmolality of original media formulations
- Manufactured in accordance with cGMP regulations
- Cell culture-tested to assure compatibility
Specifications
15° to 30°C | |
Sterile | |
100 mL |
HEPES Buffer | |
6.98 to 7.3 |