missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ BioWhittaker™ Insect Cell Culture Media: Grace's Insect Medium
Chemically-defined media optimized for specific cell types
1185.00 SEK - 1251.00 SEK
Specifications
Content And Storage | 2° to 8°C |
---|---|
Form | Liquid |
Product Type | Grace's Insect Medium |
Quantity | 500 mL |
Product Code | Brand | Product Type | Quantity | With Additives | Without Additives | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Quantity | With Additives | Without Additives | Price | Quantity & Availability | ||
11625130
|
Lonza™
04-457F |
Grace's Insect Medium | 500 mL | None | No Lactalbumin Hydrolysate, No Yeastolate, No Gentamicin |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11605140
|
Lonza™
04-649F |
Grace's Insect Medium | 500 mL | Lactalbumin Hydrolysate, Yeastolate, Gentamicin | No FBS |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Compliance
Manufactured in accordance with cGMP regulations for insect cell lines; formulations recommended by the Tissue Culture Association.
Specifications
2° to 8°C | |
Liquid | |
Grace's Insect Medium | |
500 mL |