Learn More
Lonza™ BioWhittaker™ Insect Cell Culture Media: Schneider's Drosophila Medium (Modified)
formulated for the growth of S2 cells from the fruit fly, Drosophila melanogaser. BioWhittaker™ Insect Cell Culture Media: Schneider's Drosophila Medium (Modified) may be used to culture a variety of other insect cell lines as well.
Brand: Lonza™ 04-351Q
Additional Details : Weight : 1.00000kg
Description
Used for:- Drosophila melanogaser
- Other insect cell lines
- Transient or stable transfection
- Large scale protein production in S2 cells
- malic acid
- fumaric acid
- alpha-ketoglutaric acid
- succinic acid
- sugar trehelose
- Manufactured in accordance with cGMP regulations
- The formulations used for standard liquid media products are those recommended by the Tissue Culture Association
Compliance
Manufactured in accordance with cGMP regulations for insect cell lines; formulations recommended by the Tissue Culture Association.
Specifications
2° to 8°C | |
1 L |