missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ BioWhittaker™ Phosphate Buffered Saline (10X)
Manufactured in accordance with cGMP regulations and cell culture-tested to assure compatibility
Brand: Lonza™ BE17-517Q
Additional Details : Weight : 0.50000kg
Description
- Serves as wash solution in wide variety of laboratory applications
Specifications
15° to 30°C | |
Sterile | |
7.3 to 7.5 |
Phosphate Buffered Saline (PBS) | |
No Calcium, No Magnesium | |
1 L |