Learn More
Lonza™ BioWhittaker™ PowerCHO™ CD Medium
Support the growth of CHO K1, DXB11, DG44 and GS systems
Brand: Lonza™ BE12-771Q
Additional Details : Weight : 1.00000kg
Description
PowerCHO 1, 2, and 3 Chemically Defined, Serum-free CHO Media are the next generation in CHO media, optimized for both cell growth and protein production. For therapeutic bioprocessing applications, these protein-free formulations also facilitate both downstream purification and regulatory compliance.
- Non-animal origin media
- Hydrolysate-free, serum-free
- Contains HEPES and Pluronic™ F-68
- Free of glutamine, hypoxanthine, thymidine and phenol red
- Maintain high viability (>90%) at high cell densities
- Support high-density, large scale production volumes through balanced formulation
Specifications
2° to 8°C | |
CHO | |
No Glutamine, No Phenol Red, No Hypoxanthine, No Thymidine | |
1 L |