Learn More
Lonza™ BioWhittaker™ Supplement of Insulin, Transferrin and Selenium (500X)
Manufactured in accordance with cGMP regulations
Brand: Lonza™ 17-838Z
Additional Details : Weight : 0.45360kg
Specifications
-10° to -20°C | |
Sterile | |
5 mL |
Transferrin | |
2.8 to 3.2 |