Learn More
Lonza™ BioWhittaker™ PBS EDTA
Used as a buffer in procedures generating dendritic cells from monocytes. BioWhittaker™ PBS EDTA is manufactured in accordance with cGMP regulations.
Brand: Lonza™ BE02-017F
Additional Details : Weight : 0.60000kg
Description
- For further manufacturing or laboratory use
- 500mL
Specifications
500 mL |