Learn More
Lonza™ BioWhittaker™ PBS EDTA
Used as a buffer in procedures generating dendritic cells from monocytes.  BioWhittaker™ PBS EDTA is manufactured in accordance with cGMP regulations.
Brand:  Lonza™ BE02-017F
Additional Details : Weight : 0.60000kg
Product Code. 11665100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • For further manufacturing or laboratory use
  • 500mL
Specifications
Specifications
500 mL