missing translation for 'onlineSavingsMsg'
Läs mer

Lopinavir Metabolite M-1, TRC

Produktkod. 30137174
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137174 5 mg
30009681 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137174 Supplier Toronto Research Chemicals Supplier No. TRCL4694855MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 192725-39-6
Molekylformel C37H46N4O6
Molekylvikt (g/mol) 642.78
Synonym 1(2H)-Pyrimidineacetamide, N-[(1S,3S,4S)-4-[[2-(2,6-dimethylphenoxy)acetyl]amino]-3-hydroxy-5-phenyl-1-(phenylmethyl)pentyl]tetrahydro-α-(1-methylethyl)-2,4-dioxo-, (αS)-, 1(2H)-Pyrimidineacetamide, N-[(1S,3S,4S)-4-[[(2,6-dimethylphenoxy)acetyl]amino]-3-hydroxy-5-phenyl-1-(phenylmethyl)pentyl]tetrahydro-α-(1-methylethyl)-2,4-dioxo-, (αS)- (9CI), 1(2H)-Pyrimidineacetamide, N-[4-[[(2,6-dimethylphenoxy)acetyl]amino]-3-hydroxy-5-phenyl-1-(phenylmethyl)pentyl]tetrahydro-α-(1-methylethyl)-2,4-dioxo-, [1S-[1R*(R*),3R*,4R*]]-, (αS)-N-[(1S,3S,4S)-4-[[2-(2,6-Dimethylphenoxy)acetyl]amino]-3-hydroxy-5-phenyl-1-(phenylmethyl)pentyl]tetrahydro-α-(1-methylethyl)-2,4-dioxo-1(2H)-pyrimidineacetamide
LEDER CC(C)[C@H](N1CCC(=O)NC1=O)C(=O)N[C@H](C[C@H](O)[C@H](Cc2ccccc2)NC(=O)COc3c(C)cccc3C)Cc4ccccc4

Specifications

Kemiskt namn eller material Lopinavir metabolite M-1
InChI formel InChI=1S/C37H46N4O6/c1-24(2)34(41-19-18-32(43)40-37(41)46)36(45)38-29(20-27-14-7-5-8-15-27)22-31(42)30(21-28-16-9-6-10-17-28)39-33(44)23-47-35-25(3)12-11-13-26(35)4/h5-17,24,29-31,34,42H,18-23H2,1-4H3,(H,38,45)(H,39,44)(H,40,43,46)/t29-,30-,31-,34-/m0/s1
Kvantitet 5 mg
Frakt skick Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.