Läs mer
Loracarbef Monohydrate, TRC
Produktkod. 30482333
Change view
Kvantitet:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30482333 1 mg
30481693 25 mg
30473032 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30482333 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL4695531MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 121961-22-6
Molekylformel C16 H16 Cl N3 O4 . H2 O
Molekylvikt (g/mol) 367.78
Synonym 1-Azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-chloro-8-oxo-, hydrate (1:1), (6R,7S)-, 1-Azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[(aminophenylacetyl)amino]-3-chloro-8-oxo-, monohydrate, [6R-[6α,7β(R*)]]-, 1-Azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2R)-aminophenylacetyl]amino]-3-chloro-8-oxo-, monohydrate, (6R,7S)- (9CI), LY 163892 monohydrate, Loracarbef monohydrate
IUPAC-namn (6R,7S)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-chloro-8-oxo-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate
LEDER O.N[C@@H](C(=O)N[C@H]1[C@H]2CCC(=C(N2C1=O)C(=O)O)Cl)c3ccccc3
Specifikationer
Kemiskt namn eller material Loracarbef
Kvantitet 1 mg
InChI formel InChI=1S/C16H16ClN3O4.H2O/c17-9-6-7-10-12(15(22)20(10)13(9)16(23)24)19-14(21)11(18)8-4-2-1-3-5-8;/h1-5,10-12H,6-7,18H2,(H,19,21)(H,23,24);1H2/t10-,11-,12+;/m1./s1
Formel vikt 367.0935