Läs mer
Loratadine Epoxide, TRC
Produktkod. 30476031
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30476031 100 mg
1 options
Produktkod. 30476031 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL469585100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Loratadine Epoxide
CAS 1189694-51-6
Molekylformel C22H23ClN2O3
Kvantitet 100 mg
Synonym Dispiro[11H-benzo[5,6]cyclohepta[1,2-b]pyridine-11,2'-oxirane-3',4''-piperidine]-1''-carboxylic acid, 8-chloro-5,6-dihydro-, ethyl ester, Ethyl 8-chloro-5,6-dihydrodispiro[11H-benzo[5,6]cyclohepta[1,2-b]pyridine-11,2'-oxirane-3',4''-piperidine]-1''-carboxylate, Loratadine Epoxide
InChI formel InChI=1S/C22H23ClN2O3/c1-2-27-20(26)25-12-9-21(10-13-25)22(28-21)18-8-7-17(23)14-16(18)6-5-15-4-3-11-24-19(15)22/h3-4,7-8,11,14H,2,5-6,9-10,12-13H2,1H3
LEDER CCOC(=O)N1CCC2(CC1)OC23c4ccc(Cl)cc4CCc5cccnc35
IUPAC-namn Ethyl 8-chloro-5,6-dihydrodispiro[benzo[5,6]cyclohepta[1,2-b]pyridine-11,2'-oxirane-3',4''-piperidine]-1''-carboxylate
Molekylvikt (g/mol) 398.88
Formel vikt 398.1397