Learn More
Eppendorf™ epTiPS LoRetention Dual filter Pipette tips
Optimally coordinated to Eppendorf pipettes. Eppendorf™ epTiPS offer minimal attachment and ejection forces with the highest level of tightness.
Brand:  Eppendorf™ 0030077687
Additional Details : Weight : 150.00000kg
 View more versions of this product
Product Code. 15274516
  • 59.52 SEK / 1 set
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
  • Meet EN ISO 8655 requirements
  • Ergonomically optimized cone geometry provides the optimal pipette fit on the Eppendorf pipette
  • epT.I.P.S. pipette tips can be used universally, including pipette models from other manufacturers
  • Fine graduations make visual checks of the pipetting volume even easier
  • Long, narrow tips reach the base of narrow and high vessels without touching the top of the vessel edge
  • Fine retracted or extended tips simplify work with small volumes
  • Available in purity grades of: Eppendorf Quality, PCR clean, Eppendorf Biopur™
  • Color-coded trays make volume identification and allocation of Eppendorf pipettes easier than ever
Specifications
Specifications
Low Retention
2 to 100μL
Filtered
Sample epTIPS
Videos