missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Loricrin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55439-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to Loricrin.
This gene encodes loricrin, a major protein component of the cornified cell envelope found in terminally differentiated epidermal cells. Mutations in this gene are associated with Vohwinkel′s syndrome and progressive symmetric erythrokeratoderma, both inherited skin diseases.Specifications
Loricrin | |
Polyclonal | |
Unconjugated | |
LOR | |
LOR, loricrin, LRN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16939, 4014 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.57 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P18165, P23490 | |
Lor | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |