Läs mer
Lorlatinib, MedChemExpress
Produktkod. 16477747
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
100 mg
Förpackningsstorlek:
100mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16477747 5 mg 5mg
16443286 25 mg -
16435776 100 mg 100mg
16415028 10 mg -
16401267 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16477747 Leverantör Medchem Express Leverantörsnummer HY12215
MedChemExpress Lorlatinib (PF-06463922) is a selective, orally active, brain-penetrant and ATP-competitive ROS1/ALK inhibitor. Lorlatinib has Kis of L1196M, respectively. Lorlatinib has anticancer activity.
Lorlatinib (PF-06463922) is a selective, orally active, brain-penetrant and ATP-competitive ROS1/ALK inhibitor. Lorlatinib has Kis of L1196M, respectively. Lorlatinib has anticancer activity.
Specifikationer
Kemiskt namn eller material Lorlatinib
CAS 1454846-35-5
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C21H19FN6O2
Kvantitet 5 mg
Synonym PF-06463922
Löslighetsinformation DMSO : ≥ 28 mg/mL (68.90 mM)
LEDER CN1C(C#N)=C2C(CN(C)C(C3=C([C@@H](C)OC4=C(N)N=CC2=C4)C=C(F)C=C3)=O)=N1
Molekylvikt (g/mol) 406.41
Formel vikt 406.41
Procent renhet 98.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease