Läs mer
Losartan α-Butyl-losartan Aldehyde Adduct (Losartan Impurity), TRC
Produktkod. 30005334
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005334 1 mg
30004691 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30005334 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL4705201MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1159977-48-6
Molekylformel C44 H44 Cl2 N12 O2
Molekylvikt (g/mol) 843.8
Synonym Losartan impurity B
IUPAC-namn 1-[2-butyl-5-chloro-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazol-4-yl]-2-[4-chloro-5-(hydroxymethyl)-1-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazol-2-yl]pentan-1-ol
LEDER CCCCc1nc(Cl)c(C(O)C(CCC)c2nc(Cl)c(CO)n2Cc3ccc(cc3)c4ccccc4c5nnn[nH]5)n1Cc6ccc(cc6)c7ccccc7c8nnn[nH]8
Specifikationer
Kemiskt namn eller material Losartan alpha-butyl-Losartan aldehyde adduct
Analyt- eller komponentnamn Losartan alpha-butyl-Losartan aldehyde adduct
Frakt skick Room Temperature
InChI formel InChI=1S/C44H44Cl2N12O2/c1-3-5-15-37-47-41(46)38(58(37)25-28-18-22-30(23-19-28)32-12-7-9-14-34(32)43-51-55-56-52-43)39(60)35(10-4-2)44-48-40(45)36(26-59)57(44)24-27-16-20-29(21-17-27)31-11-6-8-13-33(31)42-49-53-54-50-42/h6-9,11-14,16-23,35,39,59-60H,3-5,10,15,24-26H2,1-2H3,(H,49,50,53,54)(H,51,52,55,56)
Industry Type Forensics
Kvantitet 1 mg
Formel vikt 842.3087