Läs mer
Losartan Carboxaldehyde, TRC
Produktkod. 30467657
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30467657 100 mg
30482332 10 mg
30474049 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30467657 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL470505100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 114798-36-6
Molekylformel C22 H21 Cl N6 O
Molekylvikt (g/mol) 420.89
Synonym 2-Butyl-4-chloro-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-imidazol-5-carbaldehyde, Losartan Carboxaldehyde, Losartan Imp K (EP), Losartan Related Compound C (USP)
IUPAC-namn 2-butyl-5-chloro-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carbaldehyde
LEDER CCCCc1nc(Cl)c(C=O)n1Cc2ccc(cc2)c3ccccc3c4nnn[nH]4
Specifikationer
Kemiskt namn eller material Losartan Carboxaldehyde
Kvantitet 100 mg
InChI formel InChI=1S/C22H21ClN6O/c1-2-3-8-20-24-21(23)19(14-30)29(20)13-15-9-11-16(12-10-15)17-6-4-5-7-18(17)22-25-27-28-26-22/h4-7,9-12,14H,2-3,8,13H2,1H3,(H,25,26,27,28)
Formel vikt 420.1465